High performance foam hand wash, unfragranced, HEALTHY SOAP™ PURELL®
Leverandør: GOJO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PURELL®
115-2975EA 291 DKK
115-2975 115-2977 115-2976
High performance foam hand wash, unfragranced, HEALTHY SOAP™ PURELL®
Hudpleje Sæber og huden rensemidler
Remarkably mild soap formulation removes more than 99% of dirt and germs.
  • Formulated for dry and sensitive skin and dermatologically tested
  • Better wash experience; 95% preferred over regular soap
  • 90% naturally derived ingredients with raw materials strictly selected according to their toxicity profile
  • AT-A-GLANCE™ refills allow users to monitor product level with just one look
  • SANITARY SEALED™ PET refill is easily recycled
CLEAN RELEASE™ Technology accesses hard-to-reach areas of the skin to lift and wash away more than 99% of dirt and germs. Formulation rinses fast and clean for easy gloving, saving an average of 22 L of water per refill.