GemCell™ US origin human serum AB is xeno-free and defibrinated from source male AB plasma using therapeutic grade recombinant human thrombin.
- Recommended for process development through clinical workflows
- This product was developed without the use of animal material
- 0,1 µm sterile-filtered
GemCell™ xeno-free human serum AB is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material is defibrinated from source plasma AB. All donor units are tested for viral markers and found to be non-reactive.
Leveringsinformation: Transport temperature −70 °C.
Emballering: Shipped on dry ice.