GemCell Plus™ US origin human serum AB is defibrinated from source male AB plasma. It goes through additional rigourous viral testing.
- Recommended for process development through clincal workflows
- 0,1 µm sterile-filtered
GemCell Plus™ human serum AB is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material is defibrinated from source plasma AB. Our GemCell Plus™ goes through additional rigorous viral testing to include west Nile virus, Chagas, HTLV I/II, and HBc as well as the standard FDA required panel. All donor units are tested for viral markers and found to be non-reactive.
Leveringsinformation: Transport temperature −70 °C.
Emballering: Shipped on dry ice.