Hayward Diaphragm Valves with True Union Connections, PVC
Leverandør: Hayward
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX07390-51EA 1590 DKK
MFLX07390-51 MFLX07390-55 MFLX07390-53 MFLX07390-47 MFLX07390-49
Hayward Diaphragm Valves with True Union Connections, PVC
Ventiler Membranventiler
Designed for use in corrosive environments.
  • Multi-turn handle gives you precise control
  • Position indicator shows exact location of valve
  • Self draining allows for little dead volume in valve
  • PVC body and EPDM diaphragm
These Hayward diaphragm valves are constructed with a PVC body and EPDM diaphragm. Valves offer a maximum temperature of 140 °F (60 °C) and feature true-union connections.