Parker Hannifin Ball Valves, Polypropylene
Leverandør: Parker Hannifin
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX01379-87EA 173 DKK
MFLX01379-87 MFLX01379-65 MFLX01379-75 MFLX01379-53 MFLX01379-97 MFLX01379-67 MFLX01379-45 MFLX01379-55 MFLX01379-77 MFLX01379-61 MFLX01379-71 MFLX01379-63 MFLX01379-41 MFLX01379-73 MFLX01379-51 MFLX01379-95 MFLX01379-81 MFLX01379-69 MFLX01379-47 MFLX01379-57 MFLX01379-79 MFLX01379-49 MFLX01379-59
Parker Hannifin Ball Valves, Polypropylene
Ventiler Kugleventiler
Create a secure and easy push-to-connect connection.
  • Glass reinforced polypropylene (PP) body with a polysulfone ball and EPDM seals
  • NSF 51 listed
  • FDA compliant for contact with food
These Parker Hannifin polypropylene (PP) ball valves feature an all plastic design is leak free and corrosion resistant.