Parker Hannifin® Metal Needle Valves
Leverandør: Parker Hannifin
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX06394-09EA 1300 DKK
MFLX06394-09 MFLX06394-07 MFLX06394-15 MFLX06394-05 MFLX06394-13 MFLX06394-11
Parker Hannifin® Metal Needle Valves
Ventiler Nåleventiler
All metal needle valve features improved cycle life.
  • All metal tapered needle stem tip
  • Differential hardness between the strain hardened steam and cold formed body threads provides improved cycle life
  • Panel mountable — order panel-mounting nuts separately
These Parker Hannifin metal needle valves are designed for positive leak tight shut-off and regulation of fluids in process, power, and instrumentation applications.
Choose from 316 stainless steel (SS) or brass construction.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.