Parker Hannifin Metal Industrial Ball Valves
Leverandør: Parker Hannifin
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX30527-08EA 462 DKK
MFLX30527-08 MFLX30528-08 MFLX30527-04 MFLX30527-26 MFLX30528-04 MFLX30527-06 MFLX30527-28 MFLX30528-06 MFLX30529-06 MFLX30527-00 MFLX30528-00 MFLX30529-04 MFLX30527-02 MFLX30528-02 MFLX30529-00 MFLX30527-20
Parker Hannifin Metal Industrial Ball Valves
Ventiler Kugleventiler
Don’t worry about leaks.
  • Full flow design ensures maximum operating efficiency
  • Blow-out proof stems
Speically-designed handle increases turning leverage and provides easy opening and closing. Brass valves are an ideal choice for use in water and air service lines. Stainless steel valves are ideal for use in corrosive environments such as chemical plants, refineries, and steel mills.