Display splitter, b.safe
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BOHLR653-01EA 7270 DKK
BOHLR653-01
Display splitter, b.safe
Niveausensorer
Alarm box with aluminium case for an installation in front plates of lab furniture. Display unit to control the b.safe Splitter, which is available as an option in combination with connected fill-level sensors in b.safe Waste Caps and b.safe Waste Caps.
  • With plugs for EU, Australia, USA, Japan, and UK
  • Compatible with any version of b.safe Waste Caps
  • Additional comfort provides the possibility to snooze the alarm tone during the canister exchange
Alarm box with aluminium case for installation in the front plates of lab furniture. Front side with touch buttons and LED indicators to monitor the operation and the fill level of the connected sensors. Programmed only to control the b.safe Splitter for monitoring 2 collection containers and an automatic switching from full to empty container. Only optical alarm when reaching the nominal capacity of container 1, optical and acoustic alarm when nominal capacity of both containers is reached. The acoustic main alarm can be deactivated for 3 minutes each until an empty container is confirmed. Suitable for b.safe Waste Caps LC / System Waste Caps LC with main alarm. Includes power supply (input: AC 100 to 240 V/50 to 60 Hz, 0,5 A, output: 24 V DC, 0,5 A) with plugs for EU, Australia, USA, Japan, and UK.
Aluminium casing allows installation in the front plates of lab furniture. Easy operation via touch surface. As a special safety feature, the splitter function can use remaining volumes of full containers to prevent them from overflowing.
Information om tilbehør: Combined Signal Device with red LED flashlight and additional siren (continuous tone) can be used in a separate room or at high noise levels. Depending on the connected output socket at the b.safe Display it can be used as single alarm or to monitor several containers. IP 65-protected housing made of ABS. Includes power supply (input: AC 100 to 240 V/50 to 60 Hz, 5,5 A, output: 24 V DC, 0,5 A) and plugs for EU, Australia, USA, Japan, and UK.