Antibacterial hand wash, SURE®
Leverandør: Diversey
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SURE®
DIVY100922253EA 571 DKK
DIVY100922253 DIVY100896846 DIVY100902115
Antibacterial hand wash, SURE®
Hudpleje Cremer Hand and Body Lotions
Antibacterial hand wash ensures the highest disinfectant standards.
  • Contains plant based ingredients from renewable sources
  • 100% biodegradable and decomposed by natural processes
  • Cleans and disinfects hands in one easy step
  • Based on lactic acid, an organic based ingredient from natural sources that inhibits the growth of micro-organisms including bacteria, yeasts and some viruses
  • Passes EN test standards: EN1499, EN13727 and EN13624 (yeast only), EN1276, EN1650 EN14476 (Influenza A/H1N1) similar to the SARS-CoV-2 virus
  • Passes the latest BPR test requirements for hand disinfection
  • Triclosan and paraben free
  • No dye
  • Extensively proven skin mildness (ROAT, TEWL & Patch tests)
  • No environmental hazard classifications. No safety warnings for users
  • Bottles/cans suitable for recycling
SURE® Antibac Hand Wash is part of a unique range of plant-based, 100% biodegradable cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certificeringer: OECD 301B
Cradle to cradle certification at gold level
Emballering: 800 ml cartridge for Diversey Soft Care Line dispenser.