Du Søgte efter: Dyrefoder
17  results were found
SearchResultCount:"17"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: Thermo Scientific
Beskrivelse: Yeast-Torula
Leverandør: MP Biomedicals
Beskrivelse: An extremely fine, white, odorless tasteless, water insoluble, chemically pure cellulose powder, manufactured from natural fibres.
Leverandør: MP Biomedicals
Beskrivelse: AIN-93M powder of proteins, enzymes and peptides is part of the rodent diet.
Leverandør: MP Biomedicals
Beskrivelse: This animal diet supplement has seven-trace elements, which are usually present in a open environment - For complete description see product datasheet...
Leverandør: MP Biomedicals
Beskrivelse: Soy flour or soy grits refers to defatted soybeans ground finely.
Leverandør: MP Biomedicals
Beskrivelse: MP recommends the use of supplement U.S.P. XIII, No. 2 Mineral Mix (cat # 960300) in conjunction with this formulation for long term growth and develo...
Varenummer: (ICNA0296009801)
Leverandør: MP Biomedicals
Beskrivelse: Basic component of animal diet especially for maintaining mice and rats colonies in the research laboratory.
UOM: 1 * 1 kg
Varenummer: (ICNA0210141301)
Leverandør: MP Biomedicals
Beskrivelse: Corn-syrup is a dual acid-enzyme conversion syrup which contains approximately three times as much maltose as regular corn syrups processed to the sam...
UOM: 1 * 3,79 L
Leverandør: MP Biomedicals
Beskrivelse: This mineral mix, part of AIN-93G rodent diet, is used for growth, pregnancy and lactational phases of rodents.
Leverandør: MP Biomedicals
Beskrivelse: Corn is used as pet nutrition and natural pet food.
Varenummer: (ICNA0290283802)
Leverandør: MP Biomedicals
Beskrivelse: Hubbel, Mendel and Wakeman powder does not contain zinc, is low in manganese and phosphorus, and has a high calcium to phosphorus ratio.
UOM: 1 * 2 kg
Varenummer: (ICNA0290283410)
Leverandør: MP Biomedicals
Beskrivelse: This powdered salt mixture is used as animal diet.
UOM: 1 * 10 kg
Leverandør: MP Biomedicals
Beskrivelse: Vanderzant vitamin mixture for insects is an animal-derived component free vitamin mixture.
Varenummer: (ICNA0296040201)
Leverandør: MP Biomedicals
Beskrivelse: AIN-93-VX vitamin mixture is a component of AIN-93M and AIN-93G diet.
UOM: 1 * 1 kg
Varenummer: (ICNA0290545401)
Leverandør: MP Biomedicals
Beskrivelse: AIN-76 vitamin mixture is a basic component of animal diet for mice and rats colonies in the research lab.
UOM: 1 * 1 kg
Varenummer: (ICNA0290465401)
Leverandør: MP Biomedicals
Beskrivelse: Vitamin diet fortification mixture formulation provides sufficient vitamin levels to prevent deficiencies in all animal species, giving precise contro...
UOM: 1 * 1 kg
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 17
no targeter for Bottom