Combined refrigerator/freezer, No Frost
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
471-1159EA 0 DKK
471-1159 471-0627 471-0628
Combined refrigerator/freezer, No Frost
Køleskabe
Combined refrigerator/freezer.
  • Frost-free ventilated cooling, with two independently regulated refrigeration circuits
  • to adjust the temperature
  • in each compartment
  • White epoxy anti-corrosion treated body
  • Reversible door with depressible handle for easy opening
  • Interior lighting and automatic defrosting
  • Freezer storage in three transparent drawers for logical and easily identifiable storage
  • Devices can be fitted between two pieces furniture, adjustable feet at the front and wheels at the rear
Temperature range: +3 to +8 °C/−20 °C.