Basket sets for PG 85 glassware washers
Leverandør: Miele
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
141-1550EA 17670 DKK
141-1550 141-1551
Basket sets for PG 85 glassware washers
Opvaskemaskiner til glasvarer
Choice of Basic or Advanced basket sets for PG 85 glassware washers.
  • Suited to specific applications
  • Tailored to the needs of users
The Basic basket set is designed to accommodate both wide-necked laboratory glassware, e.g. beakers, and has an on-board spray arm. On the lower level, up to 16 narrow-necked glasses can be reprocessed using injection technology.
The Advanced basket set offers the ideal combination for large quantities of glassware as well as injection cleaning on both levels for up to 100 narrow-necked glasses. Modules are removable and can be replaced by other modules to suit the application in hand.