VariFlex
Leverandør: Agilent Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AGLS240105EA 3031 DKK
AGLS240105
VariFlex
Proteinekspressions Proteinekspressionssystemer
The VariFlex protein expression system provides vectors with unique tags that enhance the utility of E. coli as an expression host.
  • All three SET tags in every vector set
  • Gentle SBP purification tag generates high-yield, pure proteins
Leveringsinformation: Includes tag offering are three different SET tags which increase protein solubility and the SBP purification tag for high-yield, pure proteins.
Advarsel: For research use only. Not for use in diagnostic procedures.