Bond Elut™ SAX SPE cartridges
Leverandør: Agilent Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Bond Elut™
VARI22102017EA 28870 DKK
VARI22102017 VARI12102126 VARI52102017 HEWL12113056
Bond Elut™ SAX SPE cartridges
Kromatografikolonner
Bond Elut™ SAX is a strong anion-exchanger suited for extracting weak anions such as carboxylic acids that may not retain well on weak anion-exchangers.
  • Retains compounds that elute from weak anion-exchange sorbents
  • Selectivity can be user-modified for increased flexibility
  • Minimal nonpolar interactions
It is a strong anion-exchange sorbent ideally suited for the extraction of compounds, such as carboxylic acids, which may not retain effectively on weak anion-exchange sorbents.