pCMV-tag vectors
Leverandør: Agilent Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AGLS240195EA 5892 DKK
AGLS240195 AGLS240196 AGLS240197 AGLS240198 STRA240197 AGLS240203 AGLS211175 AGLS240204 AGLS240200 AGLS240202 STRA211175 STRA211174 AGLS211172 STRA211173 STRA211172 AGLS211170
pCMV-tag vectors
Vektorer, Plasmider og Biblioteker
The pCMV-Tag vectors have a single copy of the available epitope tags, while the pCMV-3Tag vectors have three copies of the epitope tag (in a variety of configurations) to prove a stronger signal in immmunological assays.
The small size of these tags ensures that they will have minimal interference with functional analysis studies.
Advarsel: For Research Use Only. Not for use in diagnostic procedures.