1,2-Dibromodifluoroethylene

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSPC9849-5GEA 6230 DKK
APOSPC9849-5G
1,2-Dibromodifluoroethylene
1,2-Dibromodifluoroethylene
Formel: C₂Br₂F₂
Kogepunkt: 71,8 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09258943
CAS-nummer: 359-21-7
UN nummer: 1993
ADR: 3,III

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR