(Trifluormethyl)trimethylsilan 99%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

APOSPC7779-100GEA 546 DKK
APOSPC7779-100G APOSPC7779-5G
(Trifluormethyl)trimethylsilan 99%
(Trifluoromethyl)trimethylsilane

Ruppert's reagent - valuable trifluoromethylating agent of electrophilic compounds.

Formel: C₄H₉F₃Si
Molekylvægt: 142,2 g/mol
Kogepunkt: 54…55 °C
Massefylde: 0,962 g/cm³ (25 °C)
Flammepunkt: –17 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00145454
CAS-nummer: 81290-20-2
UN nummer: 1993
ADR: 3,II

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR