Ethyl-3-chloro-5-(trifluoromethyl)picolinate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSPC7620-500MGEA 383 DKK
APOSPC7620-500MG
Ethyl-3-chloro-5-(trifluoromethyl)picolinate
Ethyl-3-chloro-5-(trifluoromethyl)picolinate
Formel: C₉H₇ClF₃NO₂
Molekylvægt: 253,60 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06656414
CAS-nummer: 128073-16-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR