3-Bromo-5-fluoro-1H-indazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSPC5508-250MGEA
481
DKK
APOSPC5508-250MG
3-Bromo-5-fluoro-1H-indazole
3-Bromo-5-fluoro-1H-indazole
Formel:
C₇H₄BrFN₂ Molekylvægt: 215,0200 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07781639 CAS-nummer: 885519-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...