3',5'-Dimethyl-2,2,2-trifluoroacetophenone

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSPC520513-5GEA 2630 DKK
APOSPC520513-5G
3',5'-Dimethyl-2,2,2-trifluoroacetophenone
3',5'-Dimethyl-2,2,2-trifluoroacetophenone
Formel: C₁₀H₉F₃O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01319988
CAS-nummer: 132719-10-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR