6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Farlig

APOSPC48296-25GEA 775 DKK
APOSPC48296-25G
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole 97%
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole
Formel: C₁₂H₁₃FN₂O
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03236724
CAS-nummer: 84163-77-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR