Methyl-2-fluoro-5-methylbenzoate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSPC448026-5GEA 601 DKK
APOSPC448026-5G
Methyl-2-fluoro-5-methylbenzoate
Methyl-2-fluoro-5-methylbenzoate
Formel: C₉H₉FO₂
Molekylvægt: 168,1700 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06203924
CAS-nummer: 2967-93-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR