2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSPC412228-250MGEA 688 DKK
APOSPC412228-250MG
2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 95+%
2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester
Formel: C₁₂H₁₆BF₃N₂O₂
Molekylvægt: 288,07 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12923414
CAS-nummer: 947249-01-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR