7-Fluoroquinazolin-4(3H)-one 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSPC402122-5GEA
678
DKK
APOSPC402122-5G
7-Fluoroquinazolin-4(3H)-one 97%
7-Fluoroquinazolin-4(3H)-one
Formel:
C₈H₅FN₂O Molekylvægt: 164,1400 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD05664874 CAS-nummer: 16499-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...