4-Benzyloxy-2-fluorophenylboronic acid 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSPC3973-250MGEA
110
DKK
APOSPC3973-250MG
4-Benzyloxy-2-fluorophenylboronic acid 97%
4-Benzyloxy-2-fluorophenylboronic acid
Formel:
C₁₃H₁₂BFO₃ Molekylvægt: 246,04 g/mol Smeltepunkt: 151…155 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02093064 CAS-nummer: 166744-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...