3-Fluoro-4-(phenylcarbamoyl)phenylboronic acid 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC3534-1G
APOSPC3534-1GEA
1860
DKK
APOSPC3534-1G
3-Fluoro-4-(phenylcarbamoyl)phenylboronic acid 98%
3-Fluoro-4-(phenylcarbamoyl)phenylboronic acid
Formel:
C₁₃H₁₁BFNO₃ Smeltepunkt: 219…221 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD08436025 CAS-nummer: 874288-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...