3',5'-Bis(trifluoromethyl)acetanilide 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSPC3053W-5GEA 968 DKK
APOSPC3053W-5G
3',5'-Bis(trifluoromethyl)acetanilide 97%
3',5'-Bis(trifluoromethyl)acetanilide
Formel: C₁₀H₇F₆NO
Molekylvægt: 271,16 g/mol
Smeltepunkt: 160…162 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00089454
CAS-nummer: 16143-84-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR