2-Chloro-4-(trifluoromethyl)benzyl alcohol 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSPC302909-1GEA 296 DKK
APOSPC302909-1G
2-Chloro-4-(trifluoromethyl)benzyl alcohol 98%
2-Chloro-4-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆ClF₃O
Molekylvægt: 210,58 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04972757
CAS-nummer: 56456-51-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR