2-Methyl-5-(trifluoromethoxy)benzamide 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSPC302818-1GEA
437
DKK
APOSPC302818-1G
2-Methyl-5-(trifluoromethoxy)benzamide 97%
2-Methyl-5-(trifluoromethoxy)benzamide
Formel:
C₉H₈F₃NO₂ Molekylvægt: 219,16 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD18399714 CAS-nummer: 1261828-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...