4-Methoxy-3-(trifluoromethoxy)benzonitrile 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering

APOSPC302344-1GEA
985
DKK
APOSPC302344-1G
4-Methoxy-3-(trifluoromethoxy)benzonitrile 97%
4-Methoxy-3-(trifluoromethoxy)benzonitrile
Formel:
C₉H₆F₃NO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD18398458 CAS-nummer: 1261653-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...