4'-tert-Butyl-2,2,2-trifluoroacetophenone 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Advarsel

APOSPC2501-1GEA 524 DKK
APOSPC2501-1G
4'-tert-Butyl-2,2,2-trifluoroacetophenone 97%
4'-tert-Butyl-2,2,2-trifluoroacetophenone
Formel: C₁₂H₁₃F₃O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01319979
CAS-nummer: 73471-97-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR