7-Fluorochroman-4-one 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSPC200546-5GEA
1340
DKK
APOSPC200546-5G
7-Fluorochroman-4-one 97%
7-Fluorochroman-4-one
Formel:
C₉H₇FO₂ Molekylvægt: 166,15 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06202344 CAS-nummer: 113209-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...