Methyl-3-cyano-5-fluorobenzoate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSPC200038-1GEA
470
DKK
APOSPC200038-1G
Methyl-3-cyano-5-fluorobenzoate
Methyl-3-cyano-5-fluorobenzoate
Formel:
C₉H₆FNO₂ Molekylvægt: 179,15 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD08688582 CAS-nummer: 886732-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...