3-Chloro-5-(trifluoromethyl)picolinic acid 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSPC10653-1GEA
143
DKK
APOSPC10653-1G
3-Chloro-5-(trifluoromethyl)picolinic acid 98%
3-Chloro-5-(trifluoromethyl)picolinic acid
Formel:
C₇H₃ClF₃NO₂ Molekylvægt: 225,60 g/mol Smeltepunkt: 135…138 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00277482 CAS-nummer: 80194-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...