2-Nitro-4-(trifluoromethyl)benzyl alcohol
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSPC01501-1GEA
656
DKK
APOSPC01501-1G
2-Nitro-4-(trifluoromethyl)benzyl alcohol
2-Nitro-4-(trifluoromethyl)benzyl alcohol
Formel:
C₈H₆F₃NO₃ Molekylvægt: 221,13 g/mol Smeltepunkt: 63…65 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03095418 CAS-nummer: 133605-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...