(4-Benzamidophenyl)boronic acid 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR9575-1GEA
699
DKK
APOSOR9575-1G
(4-Benzamidophenyl)boronic acid 95%
(4-Benzamidophenyl)boronic acid
Formel:
C₁₃H₁₂BNO₃ Molekylvægt: 241,05 g/mol Smeltepunkt: 194…196 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD08235089 CAS-nummer: 397843-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...