3-Cyano-5-nitrophenylboronic acid 96+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Advarsel

APOSOR9547-1GEA 2120 DKK
APOSOR9547-1G
3-Cyano-5-nitrophenylboronic acid 96+%
3-Cyano-5-nitrophenylboronic acid
Formel: C₇H₅BN₂O₄
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03411554
CAS-nummer: 913835-33-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR