3-Carboxy-4-chlorophenylboronic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR9342-1GEA
208
DKK
APOSOR9342-1G
3-Carboxy-4-chlorophenylboronic acid
3-Carboxy-4-chlorophenylboronic acid
Formel:
C₇H₆BClO₄ Molekylvægt: 200,3800 g/mol Smeltepunkt: 223…225 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD06801689 CAS-nummer: 913835-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...