Ethyl phenoxyacetate 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR925281-25GEA 524 DKK
APOSOR925281-25G
Ethyl phenoxyacetate 97%
Ethyl phenoxyacetate
Formel: C₁₀H₁₂O₃
Molekylvægt: 180,20 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00026895
CAS-nummer: 2555-49-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR