Benzylacetat 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR921864-100GEA 933 DKK
APOSOR921864-100G
Benzylacetat 95%
Benzylacetat
Formel: C₉H₁₀O₂
Molekylvægt: 150,18 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008712
CAS-nummer: 140-11-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR