3-Phenylpropionitrile 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR919888-25GEA 1260 DKK
APOSOR919888-25G
3-Phenylpropionitrile 95%
3-Phenylpropionitrile
Formel: C₉H₉N
Molekylvægt: 131,18 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001961
CAS-nummer: 645-59-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR