(Methylsulphonyl)methanesulphonyl chloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Farlig

APOSOR7845-1GEA 1320 DKK
APOSOR7845-1G
(Methylsulphonyl)methanesulphonyl chloride
(Methylsulphonyl)methanesulphonyl chloride
Formel: C₂H₅ClO₄S₂
Smeltepunkt: 110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052809
CAS-nummer: 22317-89-1
UN nummer: 3261
ADR: 8,II

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR