3-Amino-5-methylpyridazine 97+%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR61037-1GEA
2670
DKK
APOSOR61037-1G
3-Amino-5-methylpyridazine 97+%
3-Amino-5-methylpyridazine
Formel:
C₅H₇N₃ Molekylvægt: 109,1292 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD10698051 CAS-nummer: 144294-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...