4-Bromo-N-tert-butylbenzenesulphonamide 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Advarsel

APOSOR5702-5GEA 2210 DKK
APOSOR5702-5G
4-Bromo-N-tert-butylbenzenesulphonamide 98%
4-Bromo-N-tert-butylbenzenesulphonamide
Formel: C₁₀H₁₄BrNO₂S
Smeltepunkt: 112…114 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00458267
CAS-nummer: 93281-65-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR