N-Methyliminodiacetic acid 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR51834-100GEA
688
DKK
APOSOR51834-100G
N-Methyliminodiacetic acid 97%
N-Methyliminodiacetic acid
Formel:
C₅H₉NO₄ Molekylvægt: 147,13 g/mol Smeltepunkt: 220 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004284 CAS-nummer: 4408-64-4 EINECS: 224-557-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...