3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR480722-10GEA 524 DKK
APOSOR480722-10G
3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%
3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Formel: C5H4BrN5
Molekylvægt: 214,0200 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12196922
CAS-nummer: 83255-86-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR