4-Methoxythioanisole 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR480615-100GEA
1630
DKK
APOSOR480615-100G
4-Methoxythioanisole 97%
4-Methoxythioanisole
Formel:
C₈H₁₀OS Molekylvægt: 154,2294 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010587 CAS-nummer: 1879-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...