Boc-Pro-N(OMe)Me

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR451181-1GEA 2920 DKK
APOSOR451181-1G
Boc-Pro-N(OMe)Me
Boc-Pro-N(OMe)Me
Formel: C₁₂H₂₂N₂O₄
Molekylvægt: 258,3 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 115186-37-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR