5-Bromo-1-methyl-1H-indazole 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR43558-1GEA
132
DKK
APOSOR43558-1G
5-Bromo-1-methyl-1H-indazole 98%
5-Bromo-1-methyl-1H-indazole
Formel:
C₈H₇BrN₂ Molekylvægt: 211,0600 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09878568 CAS-nummer: 465529-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...