6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR41051-1GEA
492
DKK
APOSOR41051-1G
6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel:
C₈H₅ClN₂O₂ Molekylvægt: 196,59 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02186559 CAS-nummer: 182181-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...